Anti-TAS1R1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA317408
ORIGTA317408EA 680 EUR
ORIGTA317408
Anti-TAS1R1 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Immunochemistry: 6 µg/ml

Type: Primary
Antigen: TAS1R1 (taste receptor, type 1, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR