Anti-CD70 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318843
ORIGTA318843EA 680 EUR
ORIGTA318843
Anti-CD70 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 5-10 µg/ml

Type: Primary
Antigen: CD70 (CD70 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR