Anti-ID3 Mouse Monoclonal Antibody [clone: OTI4D1]

Lieferant: OriGene
TA500715
ORIGTA500715EA 616 EUR
ORIGTA500715
Anti-ID3 Mouse Monoclonal Antibody [clone: OTI4D1]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ID3 (inhibitor of DNA binding 3, dominant negative helix-loop-helix protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR