Anti-CYP2E1 Mouse Monoclonal Antibody [clone: OTI5F11]

Lieferant: OriGene
TA500717
ORIGTA500717EA 621 EUR
ORIGTA500717
Anti-CYP2E1 Mouse Monoclonal Antibody [clone: OTI5F11]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: CYP2E1 (cytochrome P450, family 2, subfamily E, polypeptide 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR