Anti-RPA2 Mouse Monoclonal Antibody [clone: OTI9A1]

Lieferant: OriGene
TA500763
ORIGTA500763EA 616 EUR
ORIGTA500763
Anti-RPA2 Mouse Monoclonal Antibody [clone: OTI9A1]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: RPA2 (replication protein A2, 32kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR