Anti-AKT2 Mouse Monoclonal Antibody [clone: OTI1A2]

Lieferant: OriGene
TA500782
ORIGTA500782EA 616 EUR
ORIGTA500782
Anti-AKT2 Mouse Monoclonal Antibody [clone: OTI1A2]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR