Anti-LEMD3 Mouse Monoclonal Antibody [clone: OTI3H2]

Lieferant: OriGene
ORIGTA500795EA 593 EUR
ORIGTA500795
Anti-LEMD3 Mouse Monoclonal Antibody [clone: OTI3H2]
Antikörper
  • Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Immunoprecipitation: 2-4μg/mg

Type: Primary
Antigen: LEMD3 (LEM domain containing 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR