Anti-PFKFB4 Mouse Monoclonal Antibody [clone: OTI1C8]

Lieferant: OriGene
TA500809
ORIGTA500809EA 621 EUR
ORIGTA500809
Anti-PFKFB4 Mouse Monoclonal Antibody [clone: OTI1C8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PFKFB4 (6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR