Anti-RALBP1 Mouse Monoclonal Antibody [clone: OTI11C6]

Lieferant: OriGene
TA500908
ORIGTA500908EA 616 EUR
ORIGTA500908
Anti-RALBP1 Mouse Monoclonal Antibody [clone: OTI11C6]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RALBP1 (ralA binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR