Anti-FAHD2A Mouse Monoclonal Antibody [clone: OTI5A7]

Lieferant: OriGene
ORIGTA500945EA 593 EUR
ORIGTA500945
Anti-FAHD2A Mouse Monoclonal Antibody [clone: OTI5A7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: FAHD2A (fumarylacetoacetate hydrolase domain containing 2A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR