Anti-MTRF1L Mouse Monoclonal Antibody [clone: OTI1D11]

Lieferant: OriGene
TA501004
ORIGTA501004EA 616 EUR
ORIGTA501004
Anti-MTRF1L Mouse Monoclonal Antibody [clone: OTI1D11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: MTRF1L (mitochondrial translational release factor 1-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR