Anti-BSG Mouse Monoclonal Antibody [clone: OTI9H5]

Lieferant: OriGene
TA501164
ORIGTA501164EA 621 EUR
ORIGTA501164
Anti-BSG Mouse Monoclonal Antibody [clone: OTI9H5]
Antikörper
  • Recommended Dilutions: Western Blot: 1:1500-3000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR