Anti-ACAT2 Mouse Monoclonal Antibody [clone: OTI1C5]

Lieferant: OriGene
TA501216
ORIGTA501216EA 616 EUR
ORIGTA501216
Anti-ACAT2 Mouse Monoclonal Antibody [clone: OTI1C5]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ACAT2 (acetyl-CoA acetyltransferase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR