Anti-TTC32 Mouse Monoclonal Antibody [clone: OTI5B4]

Lieferant: OriGene
TA501339
ORIGTA501339EA 593 EUR
ORIGTA501339
Anti-TTC32 Mouse Monoclonal Antibody [clone: OTI5B4]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: TTC32 (tetratricopeptide repeat domain 32)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR