Anti-SHPRH Mouse Monoclonal Antibody [clone: OTI4A3]

Lieferant: OriGene
TA501448
ORIGTA501448EA 593 EUR
ORIGTA501448
Anti-SHPRH Mouse Monoclonal Antibody [clone: OTI4A3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SHPRH (SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR