Anti-QPRT Mouse Monoclonal Antibody [clone: OTI4B4]

Lieferant: OriGene
ORIGTA501523EA 616 EUR
ORIGTA501523
Anti-QPRT Mouse Monoclonal Antibody [clone: OTI4B4]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: QPRT (quinolinate phosphoribosyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR