Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI7A12]

Lieferant: OriGene
TA501754
ORIGTA501754EA 593 EUR
ORIGTA501754
Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI7A12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PIK3AP1 (phosphoinositide-3-kinase adaptor protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR