Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: OTI2A2]

Lieferant: OriGene
TA501790
ORIGTA501790EA 616 EUR
ORIGTA501790
Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: OTI2A2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: EPM2AIP1 (EPM2A (laforin) interacting protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR