Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: OTI4D5]

Lieferant: OriGene
ORIGTA501854EA 616 EUR
ORIGTA501854
Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: OTI4D5]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ALDH1L1 (aldehyde dehydrogenase 1 family, member L1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR