Anti-HDAC6 Mouse Monoclonal Antibody [clone: OTI1D10]

Lieferant: OriGene
TA502000
ORIGTA502000EA 616 EUR
ORIGTA502000
Anti-HDAC6 Mouse Monoclonal Antibody [clone: OTI1D10]
Antikörper
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HDAC6 (histone deacetylase 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR