Anti-SAMHD1 Mouse Monoclonal Antibody [clone: OTI3F5]

Lieferant: OriGene
TA502024
ORIGTA502024EA 616 EUR
ORIGTA502024
Anti-SAMHD1 Mouse Monoclonal Antibody [clone: OTI3F5]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SAMHD1 (SAM domain and HD domain 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR