Anti-ANXA3 Mouse Monoclonal Antibody [clone: OTI1F11]

Lieferant: OriGene
TA502058
ORIGTA502058EA 593 EUR
ORIGTA502058
Anti-ANXA3 Mouse Monoclonal Antibody [clone: OTI1F11]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: ANXA3 (annexin A3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR