Anti-NUMB Mouse Monoclonal Antibody [clone: OTI4F3]

Lieferant: OriGene
TA502168
ORIGTA502168EA 621 EUR
ORIGTA502168
Anti-NUMB Mouse Monoclonal Antibody [clone: OTI4F3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NUMB (numb homolog (Drosophila))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR