Anti-DTNA Mouse Monoclonal Antibody [clone: OTI1B11]

Lieferant: OriGene
TA502183
ORIGTA502183EA 616 EUR
ORIGTA502183
Anti-DTNA Mouse Monoclonal Antibody [clone: OTI1B11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DTNA (dystrobrevin, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR