Anti-TRIM2 Mouse Monoclonal Antibody [clone: OTI4D6]

Lieferant: OriGene
TA502192
ORIGTA502192EA 616 EUR
ORIGTA502192
Anti-TRIM2 Mouse Monoclonal Antibody [clone: OTI4D6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TRIM2 (tripartite motif containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR