Anti-GBP5 Mouse Monoclonal Antibody [clone: OTI5C9]

Lieferant: OriGene
TA502242
ORIGTA502242EA 598 EUR
ORIGTA502242
Anti-GBP5 Mouse Monoclonal Antibody [clone: OTI5C9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GBP5 (guanylate binding protein 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR