Anti-UBL4A Mouse Monoclonal Antibody [clone: OTI2D7]

Lieferant: OriGene
ORIGTA502447EA 616 EUR
ORIGTA502447
Anti-UBL4A Mouse Monoclonal Antibody [clone: OTI2D7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: UBL4A (ubiquitin-like 4A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR