Anti-BTRC Mouse Monoclonal Antibody [clone: OTI2H2]

Lieferant: OriGene
TA502465
ORIGTA502465EA 616 EUR
ORIGTA502465
Anti-BTRC Mouse Monoclonal Antibody [clone: OTI2H2]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BTRC (beta-transducin repeat containing E3 ubiquitin protein ligase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR