Anti-RASSF5 Mouse Monoclonal Antibody [clone: OTI1H2]

Lieferant: OriGene
TA502509
ORIGTA502509EA 621 EUR
ORIGTA502509
Anti-RASSF5 Mouse Monoclonal Antibody [clone: OTI1H2]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: RASSF5 (Ras association (RalGDS/AF-6) domain family member 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR