Anti-MEF2C Mouse Monoclonal Antibody [clone: OTI2F2]

Lieferant: OriGene
ORIGTA502552EA 616 EUR
ORIGTA502552
Anti-MEF2C Mouse Monoclonal Antibody [clone: OTI2F2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR