Anti-SULT1C2 Mouse Monoclonal Antibody [clone: OTI5A11]

Lieferant: OriGene
TA502666
ORIGTA502666EA 616 EUR
ORIGTA502666
Anti-SULT1C2 Mouse Monoclonal Antibody [clone: OTI5A11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT1C2 (sulfotransferase family, cytosolic, 1C, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR