Anti-TP53 Mouse Monoclonal Antibody [clone: OTI2E4]

Lieferant: OriGene
TA502783
ORIGTA502783EA 616 EUR
ORIGTA502783
Anti-TP53 Mouse Monoclonal Antibody [clone: OTI2E4]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR