Anti-HDHD2 Mouse Monoclonal Antibody [clone: OTI3H8]

Lieferant: OriGene
TA502882
ORIGTA502882EA 616 EUR
ORIGTA502882
Anti-HDHD2 Mouse Monoclonal Antibody [clone: OTI3H8]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: HDHD2 (haloacid dehalogenase-like hydrolase domain containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR