Anti-C1orf50 Mouse Monoclonal Antibody [clone: OTI1C6]

Lieferant: OriGene
TA502937
ORIGTA502937EA 598 EUR
ORIGTA502937
Anti-C1orf50 Mouse Monoclonal Antibody [clone: OTI1C6]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C1orf50 (chromosome 1 open reading frame 50)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR