Anti-PBX1 Mouse Monoclonal Antibody [clone: OTI1H5]

Lieferant: OriGene
TA502941
ORIGTA502941EA 616 EUR
ORIGTA502941
Anti-PBX1 Mouse Monoclonal Antibody [clone: OTI1H5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PBX1 (pre-B-cell leukemia homeobox 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR