Anti-SNAP25 Mouse Monoclonal Antibody [clone: OTI4F3]

Lieferant: OriGene
TA502966
ORIGTA502966EA 616 EUR
ORIGTA502966
Anti-SNAP25 Mouse Monoclonal Antibody [clone: OTI4F3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNAP25 (synaptosomal-associated protein, 25kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR