Anti-SMS Mouse Monoclonal Antibody [clone: OTI5C12]

Lieferant: OriGene
TA503096
ORIGTA503096EA 616 EUR
ORIGTA503096
Anti-SMS Mouse Monoclonal Antibody [clone: OTI5C12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: SMS (spermine synthase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR