Anti-AK4 Mouse Monoclonal Antibody [clone: OTI1H1]

Lieferant: OriGene
TA503199
ORIGTA503199EA 616 EUR
ORIGTA503199
Anti-AK4 Mouse Monoclonal Antibody [clone: OTI1H1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AK4 (adenylate kinase 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR