Anti-PPM1B Mouse Monoclonal Antibody [clone: OTI3G9]

Lieferant: OriGene
TA503248
ORIGTA503248EA 616 EUR
ORIGTA503248
Anti-PPM1B Mouse Monoclonal Antibody [clone: OTI3G9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PPM1B (protein phosphatase, Mg2+/Mn2+ dependent, 1B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR