Anti-MSR1 Mouse Monoclonal Antibody [clone: 2G8]

Lieferant: Abnova
ABNOH00004481-M01EA 394 EUR
ABNOH00004481-M01
Anti-MSR1 Mouse Monoclonal Antibody [clone: 2G8]
Antikörper
Type: Primary
Antigen: MSR1
Clonality: Monoclonal
Clone: 2G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR