Anti-MSRA Mouse Monoclonal Antibody [clone: 3C11]

Lieferant: Abnova
ABNOH00004482-M01EA 394 EUR
ABNOH00004482-M01
Anti-MSRA Mouse Monoclonal Antibody [clone: 3C11]
Antikörper
Type: Primary
Antigen: MSRA
Clonality: Monoclonal
Clone: 3C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR