Anti-MST1R Mouse Monoclonal Antibody [clone: 3H2]

Lieferant: Abnova
ABNOH00004486-M10EA 394 EUR
ABNOH00004486-M10
Anti-MST1R Mouse Monoclonal Antibody [clone: 3H2]
Antikörper
Type: Primary
Antigen: MST1R
Clonality: Monoclonal
Clone: 3H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR