Anti-MSX1 Rabbit Polyclonal Antibody
Lieferant: Abnova
ABNOH00004487-D01EA 380 EUR
ABNOH00004487-D01
Anti-MSX1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MSX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human