Anti-MSX1 Mouse Monoclonal Antibody [clone: 2B12]

Lieferant: Abnova
ABNOH00004487-M03EA 394 EUR
ABNOH00004487-M03
Anti-MSX1 Mouse Monoclonal Antibody [clone: 2B12]
Antikörper
Type: Primary
Antigen: MSX1
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR