Anti-MSX1 Mouse Monoclonal Antibody [clone: 1E2]
Lieferant: Abnova
ABNOH00004487-M11EA 394 EUR
ABNOH00004487-M11
Anti-MSX1 Mouse Monoclonal Antibody [clone: 1E2]
Antikörper
Type: Primary
Antigen: MSX1
Clonality: Monoclonal
Clone: 1E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human