Anti-MYCN Mouse Monoclonal Antibody [clone: 4H4]
Lieferant: Abnova
ABNOH00004613-M02EA 394 EUR
ABNOH00004613-M02
Anti-MYCN Mouse Monoclonal Antibody [clone: 4H4]
Antikörper
Type: Primary
Antigen: MYCN
Clonality: Monoclonal
Clone: 4H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human