Anti-MYH7 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00004625-A01EA 258 EUR
ABNOH00004625-A01
Anti-MYH7 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MYH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human