Anti-MYL3 Mouse Monoclonal Antibody [clone: 4C2]
Lieferant: Abnova
ABNOH00004634-M12EA 394 EUR
ABNOH00004634-M12
Anti-MYL3 Mouse Monoclonal Antibody [clone: 4C2]
Antikörper
Type: Primary
Antigen: MYL3
Clonality: Monoclonal
Clone: 4C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human