Anti-MYLK Mouse Monoclonal Antibody [clone: 1D1]
Lieferant: Abnova
ABNOH00004638-M01EA 394 EUR
ABNOH00004638-M01
Anti-MYLK Mouse Monoclonal Antibody [clone: 1D1]
Antikörper
Type: Primary
Antigen: MYLK
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human