Anti-MYOC Rabbit Polyclonal Antibody
Lieferant: Abnova
ABNOH00004653-D01PEA 380 EUR
ABNOH00004653-D01P
Anti-MYOC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MYOC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human